MilliporeSigma
  • Uncaging carbon disulfide. Delivery platforms for potential pharmacological applications: a mechanistic approach.
Uncaging carbon disulfide. Delivery platforms for potential pharmacological applications: a mechanistic approach.
Chemical science (2017-10-31)
Anthony W DeMartino, Maykon Lima Souza, Peter C Ford
ABSTRACT
We describe the kinetics of the formation and decay of a series of dithiocarbamates under physiological conditions. The goal is to provide a toolbox of compounds that release CS