MilliporeSigma
  • Inhibition of the endocannabinoid-regulating enzyme monoacylglycerol lipase elicits a CB
Inhibition of the endocannabinoid-regulating enzyme monoacylglycerol lipase elicits a CB
Neuropharmacology (2017-07-05)
Robert A Owens, Mohammed A Mustafa, Bogna M Ignatowska-Jankowska, M Imad Damaj, Patrick M Beardsley, Jenny L Wiley, Micah J Niphakis, Benjamin F Cravatt, Aron H Lichtman
ABSTRACT
Substantial challenges exist for investigating the cannabinoid receptor type 1 (CB
Sigma-Aldrich
MJN110, ≥98% (HPLC)