MilliporeSigma
  • Health-promoting benefits of low-fat akawi cheese made by exopolysaccharide-producing probiotic Lactobacillus plantarum isolated from camel milk.
Health-promoting benefits of low-fat akawi cheese made by exopolysaccharide-producing probiotic Lactobacillus plantarum isolated from camel milk.
Journal of dairy science (2017-08-02)
Ayesha S Al-Dhaheri, Reem Al-Hemeiri, Jaleel Kizhakkayil, Anas Al-Nabulsi, Aisha Abushelaibi, Nagendra P Shah, Mutamed Ayyash
ABSTRACT
Lactic acid bacteria isolated from camel milk exhibit remarkable probiotic and exopolysaccharide (EPS)-producing characteristics. The health-promoting benefits of exopolysaccharide-producing probiotic Lactobacillus plantarum isolated from camel milk used for making low-fat akawi cheese were investigated. Three low-fat akawi cheeses were made using traditional culture (non-EPS-producing, EPS
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)