MilliporeSigma
  • Understanding activity trends in electrochemical water oxidation to form hydrogen peroxide.
Understanding activity trends in electrochemical water oxidation to form hydrogen peroxide.
Nature communications (2017-09-28)
Xinjian Shi, Samira Siahrostami, Guo-Ling Li, Yirui Zhang, Pongkarn Chakthranont, Felix Studt, Thomas F Jaramillo, Xiaolin Zheng, Jens K Nørskov
ABSTRACT
Electrochemical production of hydrogen peroxide (H
Sigma-Aldrich
Vanadyl acetylacetonate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Pricing and availability is not currently available.