MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Fluorescence enhancement via varied long-chain thiol stabilized gold nanoparticles: A study of far-field effect.
Fluorescence enhancement via varied long-chain thiol stabilized gold nanoparticles: A study of far-field effect.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-08-02)
Jayasmita Jana, Teresa Aditya, Mainak Ganguly, Shyamal Kumar Mehetor, Tarasankar Pal
ABSTRACT
Metal enhanced fluorescence of carbon dots has been reported in aqueous solution. Moderately fluorescing carbon dots (λ
Sigma-Aldrich
1-Octanethiol, ≥98.5%
Sigma-Aldrich
1-Tetradecanethiol, ≥98.0% (GC)
Sigma-Aldrich
Tetradecylamine, 95%