MilliporeSigma
  • Adsorption of the herbicides diquat and difenzoquat on polyurethane foam: Kinetic, equilibrium and computational studies.
Adsorption of the herbicides diquat and difenzoquat on polyurethane foam: Kinetic, equilibrium and computational studies.
Ecotoxicology and environmental safety (2017-08-13)
Jonas O Vinhal, Kassem K Nege, Mateus R Lage, José Walkimar de M Carneiro, Claudio F Lima, Ricardo J Cassella
ABSTRACT
This work reports a study about the adsorption of the herbicides diquat and difenzoquat from aqueous medium employing polyurethane foam (PUF) as the adsorbent and sodium dodecylsulfate (SDS) as the counter ion. The adsorption efficiency was shown to be dependent on the concentration of SDS in solution, since the formation of an ion-associate between cationic herbicides (diquat and difenzoquat) and anionic dodecylsulfate is a fundamental step of the process. A computational study was carried out to identify the possible structure of the ion-associates that are formed in solution. They are probably formed by three units of dodecylsulfate bound to one unit of diquat, and two units of dodecylsulfate bound to one unit of difenzoquat. The results obtained also showed that 95% of both herbicides present in 45mL of a solution containing 5.5mgL