MilliporeSigma
  • Internal Extractive Electrospray Ionization Mass Spectrometry for Quantitative Determination of Fluoroquinolones Captured by Magnetic Molecularly Imprinted Polymers from Raw Milk.
Internal Extractive Electrospray Ionization Mass Spectrometry for Quantitative Determination of Fluoroquinolones Captured by Magnetic Molecularly Imprinted Polymers from Raw Milk.
Scientific reports (2017-11-09)
Hua Zhang, Wei Kou, Aisha Bibi, Qiong Jia, Rui Su, Huanwen Chen, Keke Huang
ABSTRACT
Antibiotics contamination in food products is of increasing concern due to their potential threat on human health. Herein solid-phase extraction based on magnetic molecularly imprinted polymers coupled with internal extractive electrospray ionization mass spectrometry (MMIPs-SPE-iEESI-MS) was designed for the quantitative analysis of trace fluoroquinolones (FQs) in raw milk samples. FQs in the raw milk sample (2 mL) were selectively captured by the easily-lab-made magnetic molecularly imprinted polymers (MMIPs), and then directly eluted by 100 µL electrospraying solvent biased with +3.0 kV to produce protonated FQs ions for mass spectrometric characterization. Satisfactory analytical performance was obtained in the quantitative analysis of three kinds of FQs (i.e., norfloxacin, enoxacin, and fleroxacin). For all the samples tested, the established method showed a low limit of detection (LOD ≤ 0.03 µg L