MilliporeSigma
  • Synthesis and evaluation of antitumor, anti-inflammatory and analgesic activity of novel deoxycholic acid derivatives bearing aryl- or hetarylsulfanyl moieties at the C-3 position.
Synthesis and evaluation of antitumor, anti-inflammatory and analgesic activity of novel deoxycholic acid derivatives bearing aryl- or hetarylsulfanyl moieties at the C-3 position.
Steroids (2017-09-10)
Irina I Popadyuk, Andrey V Markov, Ekaterina A Morozova, Valeriya O Babich, Oksana V Salomatina, Evgeniya B Logashenko, Marina A Zenkova, Tat'yana G Tolstikova, Nariman F Salakhutdinov
ABSTRACT
Novel deoxycholic acid (DCA) derivatives were stereoselectively synthesised with -OH and -CH
Sigma-Aldrich
Trimethylsulfonium iodide, 98%