- Growth mechanism of strongly emitting CH
Growth mechanism of strongly emitting CH
Nature communications (2017-10-19)
He Huang, Johannes Raith, Stephen V Kershaw, Sergii Kalytchuk, Ondrej Tomanec, Lihong Jing, Andrei S Susha, Radek Zboril, Andrey L Rogach
PMID29042559
ABSTRACT
Metal halide perovskite nanocrystals are promising materials for a diverse range of applications, such as light-emitting devices and photodetectors. We demonstrate the bandgap tunability of strongly emitting CH