MilliporeSigma
  • Circulating exosomal microRNA-203 is associated with metastasis possibly via inducing tumor-associated macrophages in colorectal cancer.
Circulating exosomal microRNA-203 is associated with metastasis possibly via inducing tumor-associated macrophages in colorectal cancer.
Oncotarget (2017-11-08)
Yuki Takano, Takaaki Masuda, Hisae Iinuma, Rui Yamaguchi, Kuniaki Sato, Taro Tobo, Hidenari Hirata, Yosuke Kuroda, Sho Nambara, Naoki Hayashi, Tomohiro Iguchi, Shuhei Ito, Hidetoshi Eguchi, Takahiro Ochiya, Katsuhiko Yanaga, Satoru Miyano, Koshi Mimori
ABSTRACT
A primary tumor can create a premetastatic niche in distant organs to facilitate the development of metastasis. The mechanism by which tumor cells communicate with host cells to develop premetastatic niches is unclear. We focused on the role of microRNA (miR) signaling in promoting metastasis. Here, we identified