MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Genetic diversity of catechol 1,2-dioxygenase in the fecal microbial metagenome.
Genetic diversity of catechol 1,2-dioxygenase in the fecal microbial metagenome.
Journal of basic microbiology (2017-07-27)
Bo Xu, Caiyun Xiong, Meng Deng, Junjun Li, Xianghua Tang, Qian Wu, Junpei Zhou, Yunjuan Yang, Junmei Ding, Nanyu Han, Zunxi Huang
ABSTRACT
Catechol 1,2-dioxygenase is the key enzyme that catalyzes the cleavage of the aromatic ring of catechol. We explored the genetic diversity of catechol 1,2-dioxygenase in the fecal microbial metagenome by PCR with degenerate primers. A total of 35 gene fragments of C12O were retrieved from microbial DNA in the feces of pygmy loris. Based on phylogenetic analysis, most sequences were closely related to C12O sequences from Acinetobacter. A full-length C12O gene was directly cloned, heterologously expressed in Escherichia coli, and biochemically characterized. Purified catPL12 had optimum pH and temperature pH 8.0 and 25 °C and retained 31 and 50% of its maximum activity when assayed at 0 and 35 °C, respectively. The enzyme was stable at 25 and 37 °C, retaining 100% activity after pre-incubation for 1 h. The kinetic parameters of catPL12 were determined. The enzyme had apparent K
Sigma-Aldrich
3-Methylcatechol, 98%
Sigma-Aldrich
4-Methylcatechol, ≥95%