MilliporeSigma
  • Detection and quantification of Merkel cell polyomavirus. Analysis of Merkel cell carcinoma cases from 1977 to 2015.
Detection and quantification of Merkel cell polyomavirus. Analysis of Merkel cell carcinoma cases from 1977 to 2015.
Journal of medical virology (2017-07-07)
Marta E Álvarez-Argüelles, Santiago Melón, Susana Rojo, Ana Fernandez-Blázquez, Jose A Boga, Ana Palacio, Blanca Vivanco, María de Oña
ABSTRACT
This study investigates the presence of Merkel cell polyomavirus (MCPyV) in skin lesions of patients with Merkel cell carcinoma (MCC). MCPyV was quantified using quantitative Real-Time-PCR (qRT-PCR) in 34 paraffinized MCC samples (resected/biopsied) originally taken between 1977 and 2015, and six non-MCC samples. In 31 (91.2%) MCC-individuals, MCPyV was detected. No virus was observed in any non-MCC tumor. Average age at diagnosis was 78.2 ± 9.35 (55-97) years for women (n = 19) and 69.5 ± 14.7 (45-91) for men (n = 15) (P = 0.04). MCC tumor location, known in 25 cases, was: 11 (44%) in the head region, 6 (24%) in upper limbs, 4 (16%) in lower limbs, and 4 (16%) in the trunk. All but one patient had received some sort of treatment: 15 (45.45%) underwent both radio and chemotherapy, 13 (39.39%) only surgery, 2 (6.06%) surgery, plus radio and chemotherapy, 2 (6.06%) surgery and chemotherapy, and 1 (3.03%) only radiotherapy. Follow up data were available for 21/34 patients: recurrence was recorded for 4 (19.04%), and metastasis for 13 (61.9%). Recorded data showed that 10 men and 5 women (total 44.1%) died during follow up, 7 (46.7%) of them within 2 years of diagnosis. Viral load was 5.8 ± 1.4 log copies/10