MilliporeSigma
  • Effects of Competition, Drought Stress and Photosynthetic Productivity on the Radial Growth of White Spruce in Western Canada.
Effects of Competition, Drought Stress and Photosynthetic Productivity on the Radial Growth of White Spruce in Western Canada.
Frontiers in plant science (2017-11-23)
Syed A Alam, Jian-Guo Huang, Kenneth J Stadt, Philip G Comeau, Andria Dawson, Guillermo Gea-Izquierdo, Tuomas Aakala, Teemu Hölttä, Timo Vesala, Annikki Mäkelä, Frank Berninger
ABSTRACT
Understanding the complex interactions of competition, climate warming-induced drought stress, and photosynthetic productivity on the radial growth of trees is central to linking climate change impacts on tree growth, stand structure and in general, forest productivity. Using a mixed modeling approach, a stand-level photosynthetic production model, climate, stand competition and tree-ring data from mixedwood stands in western Canada, we investigated the radial growth response of white spruce [