MilliporeSigma
  • Calmodulin confers calcium sensitivity to the stability of the distal intracellular assembly domain of Kv7.2 channels.
Calmodulin confers calcium sensitivity to the stability of the distal intracellular assembly domain of Kv7.2 channels.
Scientific reports (2017-10-19)
Alessandro Alaimo, Eider Nuñez, Paloma Aivar, Juncal Fernández-Orth, Carolina Gomis-Perez, Ganeko Bernardo-Seisdedos, Covadonga Malo, Alvaro Villarroel
ABSTRACT
Tetrameric coiled-coil structures are present in many ion channels, often adjacent to a calmodulin (CaM) binding site, although the relationship between the two is not completely understood. Here we examine the dynamic properties of the ABCD domain located in the intracellular C-terminus of tetrameric, voltage-dependent, potassium selective Kv7.2 channels. This domain encompasses the CaM binding site formed by helices A and B, followed by helix C, which is linked to the helix D coiled-coil. The data reveals that helix D stabilizes CaM binding, promoting trans-binding (CaM embracing neighboring subunits), and they suggest that the ABCD domain can be exchanged between subunits of the tetramer. Exchange is faster when mutations in AB weaken the CaM interaction. The exchange of ABCD domains is slower in the presence of Ca2+, indicating that CaM stabilization of the tetrameric assembly is enhanced when loaded with this cation. Our observations are consistent with a model that involves a dynamic mechanism of helix D assembly, which supports reciprocal allosteric coupling between the A-B module and the coiled-coil formed by the helix D. Thus, formation of the distal helix D tetramer influences CaM binding and CaM-dependent Kv7.2 properties, whereas reciprocally, CaM and Ca2+ influence the dynamic behavior of the helix D coiled-coil.
Product Description
Millipore
Protein A-Sepharose from Staphylococcus aureus, lyophilized powder
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)