MilliporeSigma
Eldecalcitol (ED-71), an analog of 1α,25(OH)
In vitro cellular & developmental biology. Animal (2017-07-16)
T Shintani, F Takatsu, S N Z Rosli, E Usui, A Hamada, K Sumi, Y Hayashido, S Toratani, Tetsuji Okamoto
ABSTRACT
Heparin-binding protein 17 (HBp17)/fibroblast growth factor-binding protein-1 (FGFBP-1) was first purified from medium conditioned by A431 cells for its capacity to bind to fibroblast growth factors 1 and 2 (FGF-1 and -2). Among FGF family members, FGF-2 is a potent mitogen for various cell types, including vascular endothelial cells, fibroblasts, and cancer cells such as oral squamous cell carcinoma (OSCC) cells. Besides being well known in bone metabolism, the active form of vitamin D
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O