MilliporeSigma
  • Building on Sub-Arctic Soil: Geopolymerization of Muskeg to a Densified Load-Bearing Composite.
Building on Sub-Arctic Soil: Geopolymerization of Muskeg to a Densified Load-Bearing Composite.
Scientific reports (2017-11-09)
Gregory R Waetzig, Junsang Cho, Max Lacroix, Sarbajit Banerjee
ABSTRACT
The marshy water-saturated soil typical of the sub-Arctic represents a considerable impediment to the construction of roads, thereby greatly hindering human habitation and geological excavation. Muskeg, the native water-laden topsoil characteristic of the North American sub-Arctic, represents a particularly vexing challenge for road construction. Muskeg must either be entirely excavated, or for direct construction on muskeg, a mix of partial excavation and gradual compaction with the strategic placement of filling materials must be performed. Here, we demonstrate a novel and entirely reversible geopolymerization method for reinforcing muskeg with wood fibers derived from native vegetation with the addition of inorganic silicate precursors and without the addition of extraneous metal precursors. A continuous siloxane network is formed that links together the muskeg, wood fibers, and added silicates yielding a load-bearing and low-subsidence composite. The geopolymerization approach developed here, based on catalyzed formation of a siloxane network with further incorporation of cellulose, allows for an increase of density as well as compressive strength while reducing the compressibility of the composite.