MilliporeSigma
  • Retinoic acid induction of CD1d expression primes chronic lymphocytic leukemia B cells for killing by CD8
Retinoic acid induction of CD1d expression primes chronic lymphocytic leukemia B cells for killing by CD8
Clinical immunology (Orlando, Fla.) (2017-08-07)
Yasmeen G Ghnewa, Vincent P O'Reilly, Elisabeth Vandenberghe, Paul V Browne, Anthony M McElligott, Derek G Doherty
ABSTRACT
Invariant natural killer T (iNKT) cells are cytotoxic T cells that respond to glycolipid antigens presented by CD1d. Therapeutic activation of iNKT cells with α-galactosylceramide (α-GalCer) can prevent and reverse tumor growth in mice and clinical trials involving α-GalCer-stimulated iNKT cells are ongoing in humans. B cells express CD1d, however, we show that CD1d expression is reduced on B cells from patients with chronic lymphocytic leukemia (CLL). B cells from CLL patients pulsed with α-GalCer failed to stimulate cytolytic degranulation by iNKT cell lines, but could present the more potent glycolipid analogue, 7DW8-5. Retinoic acid receptor-α (RAR-α) agonists induced CD1d expression by CLL B cells, restoring their ability to present α-GalCer to CD8α
Sigma-Aldrich
AM580, ≥98% (HPLC)
Pricing and availability is not currently available.