MilliporeSigma
  • Effects of cyclosporin A and glucocorticosteroids on antigen-induced hypersensitivity to histamine in a guinea pig model of allergic rhinitis.
Effects of cyclosporin A and glucocorticosteroids on antigen-induced hypersensitivity to histamine in a guinea pig model of allergic rhinitis.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (1998-04-16)
S I Narita, K Asakura, H Shiraski, M Isobe, H Ogasawara, H Saito, A Kataura
ABSTRACT
In an attempt to study the pathogenesis of mucosal hypersensitivity in allergic rhinitis, we investigated the suppressive effects of cyclosporin A (CyA) and glucocorticosteroids on ovalbumin (OA)-induced hypersensitivity to topical histamine challenge. Actively sensitized Dunkin-Hartley guinea pigs. OA and alum were applied to guinea pigs intraperitoneally 3 times at two-week intervals. After general sensitization, OA inhalation was performed every day for 6 days as topical sensitization. Before inhalation, treatment with CyA (50 mg/kg, p.o.), glucocorticosteroids (beclomethasone propionate (1.0 mg/kg, i.p.), fluticasone propionate (FP, 0.5 mg/kg, i.p.)) or vehicle were performed, and the sensitivity to histamine was measured before and after the inhalation. Moreover, in actively (general and topical) sensitized guinea pigs, FP (0.5 mg/kg, i.p.) was applied every day for 5 days and histamine sensitivity was evaluated before and after the application. We found that histamine sensitivity was significantly increased by nasal antigen challenge in this guinea pig model, and that the occurrence of histamine hypersensitivity was inhibited by the pretreatment with CyA and glucocorticosteroids. Although multiple administration of FP gradually reduced the histamine hypersensitivity according to the period of administration, it did not significantly alter the histamine hypersensitivity after the occurrence of hypersensitivity. It is concluded that CyA and glucocorticosteroids suppress antigen-induced histamine hypersensitivity in a guinea pig model of allergic rhinitis.
Sigma-Aldrich
Beclomethasone, ≥99%