MilliporeSigma
  • Effect of ginkgo extract supplementation on in vitro rumen fermentation and bacterial profiles under different dietary conditions.
Effect of ginkgo extract supplementation on in vitro rumen fermentation and bacterial profiles under different dietary conditions.
Animal science journal = Nihon chikusan Gakkaiho (2017-07-15)
Seongjin Oh, Satoshi Koike, Yasuo Kobayashi
ABSTRACT
Ginkgo extract was applied to a batch culture study and evaluated for its potential as a feed additive for ruminant animals under different forage-to-concentrate (F:C) ratios (1:9, 3:7, 5:5, 7:3 and 9:1). Rumen fluid was mixed with respective diet and incubated at 39°C for 24 h with and without ginkgo extract (1.6% fruit equivalent in culture). Methane production was significantly decreased by ginkgo extract, with the greatest reductions found in the 5:5 (41.9%) followed by the 7:3 ratios (36.7%). Total short chain fatty acid and ammonia levels were not affected by ginkgo extract supplementation in any of the five different diets. However, ginkgo extract increased propionate proportion and decreased acetate proportion in all dietary conditions tested. The levels of total bacteria, Ruminococcus flavefaciens, Ruminococcus albus and Fibrobacter succinogenes were decreased by ginkgo extract. The levels of Selenomonas ruminantium, Anaerovibrio lipolytica, Ruminobacter amylophilus, Succinivibrio dextrinosolvens and Megasphaera elsdenii were increased by ginkgo extract supplementation, possibly contributing to the higher propionate production. These results suggest that rumen modulation by ginkgo extract can be achieved at a wide range of F:C ratios with no adverse impact on feed digestion. Moreover, F:C ratios of 5:5 and 7:3 may be optimal when methane mitigation is expected.
Product Description
Supelco
5-Pentadecylresorcinol, analytical standard