MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Lethality of inappropriate plasma exposure on chicken embryonic development.
Lethality of inappropriate plasma exposure on chicken embryonic development.
Oncotarget (2017-11-22)
Jiao Jiao Zhang, Jin Oh Jo, Do Luong Huynh, Mrinmoy Ghosh, Nameun Kim, Sang Baek Lee, Hak Kyo Lee, Young Sun Mok, Taeho Kwon, Dong Kee Jeong
ABSTRACT
In this study, we examined the effects of non-thermal dielectric barrier discharge plasma on embryonic development in chicken eggs in order to determine the optimal level of plasma exposure for the promotion of embryonic growth. We exposed developing chicken embryos at either Hamburger-Hamilton (HH) stage 04 or HH 20 to plasma at voltages of 11.7 kV to 27.6 kV. Our results show exposure at 11.7 kV for 1 min promoted chicken embryonic development, but exposure to more duration and intensity of plasma resulted in dose-dependent embryonic death and HH 20 stage embryos survive longer than those at stage HH 04. Furthermore, plasma exposure for 4 min increased the production of reactive oxygen species (ROS) and inactivated the nuclear factor erythroid 2-related factor 2 (NRF2)-antioxidant response signaling pathway, resulting in suppression of antioxidant enzymes in the skeletal muscle tissue of the dead embryos. We also found decreased levels of adenosine triphosphate production and reductions in the expression levels of several growth-related genes and proteins. These findings indicate that inappropriate plasma exposure causes dose-dependent embryonic death via excessive accumulation of ROS, NRF2-antioxidant signaling pathway disruption, and decreased growth factor expression.
Sigma-Aldrich
NSC 632839, ≥98% (HPLC)
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)