MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Characterization of muscarinic receptors in rat kidney.
European journal of pharmacology (1993-01-04)
W M Blankesteijn, H L Siero, J F Rodrigues de Miranda, Y J van Megen, F G Russel
ABSTRACT
Muscarinic receptors in mammalian kidney seem to be involved in diuresis. In this study we give a detailed characterization of receptors in rat kidney. Specific binding of [3H](-)-quinuclidinylbenzilate ([3H]QNB) to membranes of rat kidney cortex was saturable and of high affinity. A dissociation constant of 0.063 +/- 0.003 nM and a receptor density of 1.46 +/- 0.07 pmol/g wet weight were obtained. The dissociation kinetics could be best described by assuming a mono-exponential function (k-1 = (0.52 +/- 0.1) x 10(-4) s-1). The binding of [3H]QNB reached a maximum in 60 min at 0.6 nM at 37 degrees C. Competition experiments with the enantiomers of benzetimide confirmed the muscarinic nature of the [3H]QNB binding sites. The inhibition constants of pirenzepine (0.23 +/- 0.02 microM), (+-)-hexahydrosiladifenidol (0.040 +/- 0.002 microM), AF-DX 116 (1.45 +/- 0.07 microM), methoctramine (1.67 +/- 0.02 microM) and gallamine (78 +/- 3 microM) classified this receptor as an M3 receptor. Inhibition of [3H]QNB binding by the agonists methylfurtrethonium, arecoline, isoarecoline methiodide, arecaidine propargyl ester and McN-A-343 displayed monophasic inhibition curves. With (+/-)-cis-2-methyl-4-dimethylaminomethyl-1,3- dioxolane methiodide in two out of four experiments a small (11%) population of high affinity agonist sites could be detected. The potassium sparing diuretic amiloride inhibited [3H]QNB binding (36 +/- 3 microM). Although in a way related to the amiloride binding site, the muscarinic receptors in rat kidney are unlikely to be the primary target of diuretic action of this drug.