MilliporeSigma
T-Cell Mediation of Pregnancy Analgesia Affecting Chronic Pain in Mice.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-09-08)
Sarah F Rosen, Boram Ham, Shannon Drouin, Nadia Boachie, Anne-Julie Chabot-Dore, Jean-Sebastien Austin, Luda Diatchenko, Jeffrey S Mogil
ABSTRACT
It has been reported consistently that many female chronic pain sufferers have an attenuation of symptoms during pregnancy. Rats display increased pain tolerance during pregnancy due to an increase in opioid receptors in the spinal cord. Past studies did not consider the role of non-neuronal cells, which are now known to play an important role in chronic pain processing. Using an inflammatory (complete Freund's adjuvant) or neuropathic (spared nerve injury) model of persistent pain, we observed that young adult female mice in early pregnancy switch from a microglia-independent to a microglia-dependent pain hypersensitivity mechanism. During late pregnancy, female mice show no evidence of chronic pain whatsoever. This pregnancy-related analgesia is reversible by intrathecal administration of naloxone, suggesting an opioid-mediated mechanism; pharmacological and genetic data suggest the importance of δ-opioid receptors. We also observe that T-cell-deficient (
Sigma-Aldrich
Cell dissociation sieve - tissue grinder kit, tissue grinder homogenizer kit, stainless steel, autoclavable, 1/kit
Pricing and availability is not currently available.