MilliporeSigma
  • Determination of acyclovir in human serum by high-performance liquid chromatography using liquid?liquid extraction and its application in pharmacokinetic studies
Determination of acyclovir in human serum by high-performance liquid chromatography using liquid?liquid extraction and its application in pharmacokinetic studies
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences (2005)
Bahrami.Gh, et al.