MilliporeSigma
  • The effects of processing methods on the levels of lysine, methionine and the general acceptability of ogi processed using starter cultures
The effects of processing methods on the levels of lysine, methionine and the general acceptability of ogi processed using starter cultures
International Journal of Food Microbiology (2001)
O.D.Teniola and S.A.Odunfa