MilliporeSigma
  • Identification of bacteria potentially producing biogenic amines isolated from Venezuelan white cheese
Identification of bacteria potentially producing biogenic amines isolated from Venezuelan white cheese
Revista Brasileira de Farmacognosia (2016)
Alba Lorena BENAVIDES Sierra