MilliporeSigma
  • Sequential Vacc-4x and romidepsin during combination antiretroviral therapy (cART): Immune responses to Vacc-4x regions on p24 and changes in HIV reservoirs.
Sequential Vacc-4x and romidepsin during combination antiretroviral therapy (cART): Immune responses to Vacc-4x regions on p24 and changes in HIV reservoirs.
The Journal of infection (2017-09-18)
G Tapia, J F Højen, M Ökvist, R Olesen, S Leth, S K Nissen, D J VanBelzen, U O'Doherty, A Mørk, K Krogsgaard, O S Søgaard, L Østergaard, M Tolstrup, G Pantaleo, M A Sommerfelt
ABSTRACT
The REDUC clinical study Part B investigated Vacc-4x/rhuGM-CSF therapeutic vaccination prior to HIV latency reversal using romidepsin. The main finding was a statistically significant reduction from baseline in viral reservoir measurements. Here we evaluated HIV-specific functional T-cell responses following Vacc-4x/rhuGM-CSF immunotherapy in relation to virological outcomes on the HIV reservoir. This study, conducted in Aarhus, Denmark, enrolled participants (n = 20) with CD4>500 cells/mm The frequency of participants with CD8+ T-cell proliferation assay positivity was 8/16 (50%) at baseline, 11/15 (73%) post-vaccination, 6/14 (43%) during romidepsin, and 9/15 (60%)post-romidepsin. Participants with CD8+ T-cell proliferation assay positivity post-vaccination showed reductions in total HIV DNA post-vaccination (p = 0.006; q = 0.183), post-latency reversal (p = 0.005; q = 0.183), and CA-RNA reductions post-vaccination (p = 0.015; q = 0.254). Participants (40%) were defined as proliferation 'Responders' having ≥2-fold increase in assay positivity post-baseline. Robust ELISpot baseline responses were found in 87.5% participants. No significant changes were observed in the proportion of polyfunctional CD8+ T-cells to HIV In this 'shock and kill' approach supported by therapeutic vaccination, CD8+ T-cell proliferation represents a valuable means to monitor functional immune responses as part of the path towards functional HIV cure.
Sigma-Aldrich
Romidepsin, ≥98% (HPLC)