MilliporeSigma
Monomeric M2e antigen in VesiVax
Journal of liposome research (2017-09-19)
J P Adler-Moore, W Ernst, H Kim, N Ward, S M Chiang, T Do, G Fujii
ABSTRACT
Given the interest in the ectodomain of the matrix 2 (M2e) channel protein as a target for development of a universal influenza vaccine, we examined the role of the antigen configuration of M2e in generating a protective immune response. A series of M2e mutations and a truncated M2e segment were prepared as a means of controlling the formation of monomer, dimer, and higher order multimeric forms of M2e. Each of these M2e peptides was incorporated into a liposome-based vaccine technology platform previously shown to stimulate a protective response to influenza A infection using M2e as a mixture of monomers, dimers and multimers (L-M2e1-HD/MPL). Our results using these modified forms of M2e produced 90-100% survival following lethal challenge with H1N1 (A/PR/8/34) in both inbred BALB/c and outbred Swiss Webster mice vaccinated with a truncated monomeric form of the M2 protein, M2e
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)