MilliporeSigma
Adenosine A
American journal of physiology. Renal physiology (2017-09-01)
Hitesh Soni, Dieniffer Peixoto-Neves, Randal K Buddington, Adebowale Adebiyi
ABSTRACT
Adenosine, a regulator of cardiovascular development and renal function, constricts renal afferent arterioles by inducing intracellular Ca
Sigma-Aldrich
Anti-ADORA1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
2-Chloro-N6-cyclopentyladenosine, adenosine receptor agonist