MilliporeSigma
  • Dual-wavelength light-emitting diode-based ultraviolet absorption detector for nano-flow capillary liquid chromatography.
Dual-wavelength light-emitting diode-based ultraviolet absorption detector for nano-flow capillary liquid chromatography.
Journal of chromatography. A (2017-08-19)
Xiaofeng Xie, Luke T Tolley, Thy X Truong, H Dennis Tolley, Paul B Farnsworth, Milton L Lee
ABSTRACT
The design of a miniaturized LED-based UV-absorption detector was significantly improved for on-column nanoflow LC. The detector measures approximately 27mm×24mm×10mm and weighs only 30g. Detection limits down to the nanomolar range and linearity across 3 orders of magnitude were obtained using sodium anthraquinone-2-sulfonate as a test analyte. Using two miniaturized detectors, a dual-detector system was assembled containing 255nm and 275nm LEDs with only 216nL volume between the detectors A 100μm slit was used for on-column detection with a 150μm i.d. packed capillary column. Chromatographic separation of a phenol mixture was demonstrated using the dual-detector system, with each detector producing a unique chromatogram. Less than 6% variation in the ratios of absorbances measured at the two wavelengths for specific analytes was obtained across 3 orders of magnitude concentration, which demonstrates the potential of using absorption ratio measurements for target analyte detection. The dual-detector system was used for simple, but accurate, mobile phase flow rate measurement at the exit of the column. With a flow rate range from 200 to 2000nL/min, less than 3% variation was observed.
Sigma-Aldrich
Sodium anthraquinone-2-sulfonate, ≥98% (HPLC)
Pricing and availability is not currently available.