MilliporeSigma
  • Anthrax lethal toxin (LeTx) neutralization by PA domain specific antisera.
Anthrax lethal toxin (LeTx) neutralization by PA domain specific antisera.
Toxicon : official journal of the International Society on Toxinology (2017-09-19)
Monika Verma, Nagendra Suryanarayana, Urmil Tuteja, Kulanthaivel Thavachelvam, M K Rao, Rakesh Bhargava, Sangeeta Shukla
ABSTRACT
Anthrax associated causalities in humans and animals are implicated mainly due to the action of two exotoxins that are secreted by the bacterium Bacillus antharcis during the infection. These exotoxins comprise of three protein components namely protective antigen (PA), lethal factor (LF) and edema factor (EF). The protective antigen is the common toxin component required to form both lethal toxin (LeTx) and edema toxin (EdTx). The LeTx is formed, when PA combines with LF and EdTx is formed when PA combines with EF. Therapeutic interventions aiming to neutralize these key effectors of anthrax pathology would therefore, provide an effective means to counter the toxicity imposed by the anthrax toxins on the host. The present work describes the lethal toxin neutralization potential of polyclonal antisera developed against the individual domains of the protective antigen component of the anthrax toxin. The individual domains were produced as recombinant proteins in E. coli and validated with peptide mass fingerprinting by MALDI-TOF analysis and corresponding mice polyclonal antisera by western blotting. Each domain specific antibody titre and isotype was ascertained by ELISA. The isotyping revealed the predominance of IgG1 isotype. The toxin neutralizing potential of these domain specific antisera were evaluated by in-vitro cell viability MTT assay, employing J774.1 mouse macrophage cell line against LeTx (0.25 μg ml
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 15 mL
Pricing and availability is not currently available.