MilliporeSigma
  • Drug repurposing strategy against Trypanosoma cruzi infection: In vitro and in vivo assessment of the activity of metronidazole in mono- and combined therapy.
Drug repurposing strategy against Trypanosoma cruzi infection: In vitro and in vivo assessment of the activity of metronidazole in mono- and combined therapy.
Biochemical pharmacology (2017-09-06)
M R Simões-Silva, J S De Araújo, G M Oliveira, K C Demarque, R B Peres, I D'Almeida-Melo, D G J Batista, C F Da Silva, C Cardoso-Santos, P B Da Silva, M M Batista, M T Bahia, M N C Soeiro
ABSTRACT
Metronidazole (Mtz) is a commercial broad-spectrum nitroimidazolic derivative with relevant antimicrobial activity and relative safety profile. Therefore, it is fair to consider Mtz a candidate for drug repurposing for other neglected conditions such as Chagas disease (CD), a parasitic pathology caused by Trypanosoma cruzi. CD is treated only with benznidazole (Bz) and nifurtimox, both introduced in clinics decades ago despite important limitations, including low efficacy on the later disease stage (chronic form) and severe side effects. New cheap and fast alternative treatments for CD are needed, thus the repurposing of Mtz was assessed in vitro and in vivo in mono- and combined therapy. In vitro assays demonstrated EC
Sigma-Aldrich
Chlorophenol Red, indicator grade