MilliporeSigma
  • Exploring the Anti-quorum Sensing and Antibiofilm Efficacy of Phytol against
Exploring the Anti-quorum Sensing and Antibiofilm Efficacy of Phytol against
Frontiers in cellular and infection microbiology (2017-12-21)
Ramanathan Srinivasan, Ramar Mohankumar, Arunachalam Kannappan, Veeramani Karthick Raja, Govindaraju Archunan, Shunmugiah Karutha Pandian, Kandasamy Ruckmani, Arumugam Veera Ravi
ABSTRACT
Quorum Sensing (QS) mechanism, a bacterial density-dependent gene expression system, governs the