MilliporeSigma
  • CHROMagar COL-APSE: a selective bacterial culture medium for the isolation and differentiation of colistin-resistant Gram-negative pathogens.
CHROMagar COL-APSE: a selective bacterial culture medium for the isolation and differentiation of colistin-resistant Gram-negative pathogens.
Journal of medical microbiology (2017-10-07)
Muhd Haziq F Abdul Momin, David C Bean, Rene S Hendriksen, Marisa Haenni, Lynette M Phee, David W Wareham
ABSTRACT
A selective chromogenic culture medium for the laboratory isolation and differentiation of colistin resistant Acinetobacter, Pseudomonas, Stenotrophomonas and Enterobacteriaceae spp. (CHROMagar COL-APSE) was developed, evaluated and compared to an existing selective bacterial culture medium (SuperPolymyxin). The medium was challenged with 84 isolates, including polymyxin B (POL B)-susceptible and -resistant type strains and colistin (COL)-resistant organisms recovered from human and animal samples. Susceptibility to COL and POL B was determined by agar dilution and broth microtitre dilution. The lower limit for the detection of COL-resistant organisms was also calculated for both CHROMagar COL-APSE and SuperPolymyxin media. The ability to isolate and correctly differentiate COL-resistant organisms within mixed cultures was also assessed and compared using both media. Using CHROMagar COL-APSE, Gram-negative pathogens (n=71) with intrinsic (n=8) or acquired COL (n=63) resistance were recovered with 100 % specificity down to the lower limit of detection of 10 CHROMagar COL-APSE is a sensitive and specific medium for the growth of COL-resistant bacterial pathogens. Due to the low limit of detection (10
Sigma-Aldrich
Daptomycin, cyclic lipopeptide antibiotic