MilliporeSigma
  • Changes in the inspiratory pattern contribute to modulate the sympathetic activity in sino-aortic denervated rats.
Changes in the inspiratory pattern contribute to modulate the sympathetic activity in sino-aortic denervated rats.
Experimental physiology (2017-06-24)
Mateus R Amorim, Leni G H Bonagamba, George M P R Souza, Davi J A Moraes, Benedito H Machado
ABSTRACT
What is the central question of this study? Sino-aortic denervated (SAD) rats present normal levels of sympathetic activity and mean arterial pressure. However, neural mechanisms regulating the sympathetic activity in the absence of arterial baroreceptors remain unclear. Considering that respiration modulates the sympathetic activity, we hypothesize that changes in the respiratory network contribute to keep the sympathetic outflow in the normal range after removal of arterial baroreceptors. What is the main finding and its importance? Despite longer inspiration observed in SAD rats, the respiratory-sympathetic coupling is working within a normal range of variation. These findings suggest that in the absence of arterial baroreceptors the respiratory modulation of sympathetic activity is maintained within the normal range. The activity of presympathetic neurons is under respiratory modulation, and changes in the central respiratory network may impact on the baseline sympathetic activity and mean arterial pressure. It is well known that after removal of baroreceptor afferents [sino-aortic denervation (SAD)], rats present an unexpected normal level of mean arterial pressure. We hypothesized that changes in the respiratory pattern and in the respiratory modulation of the sympathetic activity contribute to keep the sympathetic outflow within a normal range of variation in the absence of arterial baroreceptors in rats. To study these mechanisms, we recorded perfusion pressure and the activities of phrenic and thoracic sympathetic nerves in male juvenile rats using the working heart-brainstem preparation. The time of inspiration significantly increased in SAD rats, and this change was not dependent on the carotid bodies or on the vagal afferents. However, no changes were observed in the perfusion pressure or in the baseline thoracic sympathetic nerves in all phases of the respiratory cycle in SAD rats. Our data show that despite longer inspiratory activity, the baseline sympathetic activity is maintained at a normal level in SAD rats. These findings indicate that the respiratory-sympathetic coupling is normal after SAD and suggest that the respiratory modulation of sympathetic activity is maintained within the normal range after the removal of arterial baroreceptors.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Supelco
Flunixin, VETRANAL®, analytical standard