MilliporeSigma
A scale to measure MRI contrast agent sensitivity.
Scientific reports (2017-11-16)
Rohan D A Alvares, Daniel A Szulc, Hai-Ling M Cheng
ABSTRACT
Magnetic resonance imaging (MRI) provides superior resolution of anatomical features and the best soft tissue contrast, and is one of the predominant imaging modalities. With this technique, contrast agents are often used to aid discrimination by enhancing specific features. Over the years, a rich diversity of such agents has evolved and with that, so has a need to systematically sort contrast agents based on their efficiency, which directly determines sensitivity. Herein, we present a scale to rank MRI contrast agents. The scale is based on analytically determining the minimum detectable concentration of a contrast agent, and employing a ratiometric approach to standardize contrast efficiency to a benchmark contrast agent. We demonstrate the approach using several model contrast agents and compare the relative sensitivity of these agents for the first time. As the first universal metric of contrast agent sensitivity, this scale will be vital to easily assessing contrast agent efficiency and thus important to promoting use of some of the elegant and diverse contrast agents in research and clinical practice.