MilliporeSigma
  • The Traditional Japanese Herbal Medicine Hachimijiogan Elicits Neurite Outgrowth Effects in PC12 Cells and Improves Cognitive in AD Model Rats via Phosphorylation of CREB.
The Traditional Japanese Herbal Medicine Hachimijiogan Elicits Neurite Outgrowth Effects in PC12 Cells and Improves Cognitive in AD Model Rats via Phosphorylation of CREB.
Frontiers in pharmacology (2017-12-07)
Kaori Kubota, Haruka Fukue, Hitomi Sato, Kana Hashimoto, Aya Fujikane, Hiroshi Moriyama, Takuya Watanabe, Shutaro Katsurabayashi, Mosaburo Kainuma, Katsunori Iwasaki
ABSTRACT
Hachimijiogan (HJG) is a traditional herbal medicine that improves anxiety disorders in patients with dementia. In this study, we tested the hypothesis that HJG exerts neurotrophic factor-like effects to ameliorate memory impairment in Alzheimer disease (AD) model rats. First, we describe that HJG acts to induce neurite outgrowth in PC12 cells (a rat pheochromocytoma cell line) like nerve growth factor (NGF) in a concentration-dependent manner (3 μg/ml HJG,
Sigma-Aldrich
Naphthol AS-E phosphate, suitable for histology