MilliporeSigma
A Microdevice Platform Recapitulating Hypoxic Tumor Microenvironments.
Scientific reports (2017-11-11)
Yuta Ando, Hoang P Ta, Daniel P Yen, Sang-Sin Lee, Sneha Raola, Keyue Shen
ABSTRACT
Hypoxia plays a central role in cancer progression and resistance to therapy. We have engineered a microdevice platform to recapitulate the intratumor oxygen gradients that drive the heterogeneous hypoxic landscapes in solid tumors. Our design features a "tumor section"-like culture by incorporating a cell layer between two diffusion barriers, where an oxygen gradient is established by cellular metabolism and physical constraints. We confirmed the oxygen gradient by numerical simulation and imaging-based oxygen sensor measurement. We also demonstrated spatially-resolved hypoxic signaling in cancer cells through immunostaining, gene expression assay, and hypoxia-targeted drug treatment. Our platform can accurately generate and control oxygen gradients, eliminates complex microfluidic handling, allows for incorporation of additional tumor components, and is compatible with high-content imaging and high-throughput applications. It is well suited for understanding hypoxia-mediated mechanisms in cancer disease and other biological processes, and discovery of new therapeutics.
Sigma-Aldrich
Pimonidazole, ≥98% (HPLC)