MilliporeSigma
Ginsenoside Rk1 bioactivity: a systematic review.
PeerJ (2017-11-22)
Abdelrahman Elshafay, Ngo Xuan Tinh, Samar Salman, Yara Saber Shaheen, Eman Bashir Othman, Mohamed Tamer Elhady, Aswin Ratna Kansakar, Linh Tran, Le Van, Kenji Hirayama, Nguyen Tien Huy
ABSTRACT
Ginsenoside Rk1 (G-Rk1) is a unique component created by processing the ginseng plant (mainly Sung Ginseng (SG)) at high temperatures. The aim of our study was to systematically review the pharmacological effects of G-Rk1. We utilized and manually searched eight databases to select