MilliporeSigma
  • miR-217 regulates tumor growth and apoptosis by targeting the MAPK signaling pathway in colorectal cancer
miR-217 regulates tumor growth and apoptosis by targeting the MAPK signaling pathway in colorectal cancer
Oncology Letters (2016)
Zhang N, et al.