MilliporeSigma
  • High-yield synthesis of single-crystalline antimony telluride hexagonal nanoplates using a solvothermal approach.
High-yield synthesis of single-crystalline antimony telluride hexagonal nanoplates using a solvothermal approach.
Journal of the American Chemical Society (2005-10-06)
Wenzhong Wang, Bed Poudel, Jian Yang, D Z Wang, Z F Ren
ABSTRACT
We report a high-yield synthesis of single-crystal hexagonal Sb2Te3 nanoplates using a solvothermal approach. The experimental results show that the concentration of capping agent CTAB played a key role in the formation of nanoplates. This new nanostructure may provide opportunity for the exploration of novel thermoelectric properties and have potential applications in thermoelectric nanodevices.