MilliporeSigma
  • A potent and selective S1P(1) antagonist with efficacy in experimental autoimmune encephalomyelitis.
A potent and selective S1P(1) antagonist with efficacy in experimental autoimmune encephalomyelitis.
Chemistry & biology (2012-09-25)
Jean Quancard, Birgit Bollbuck, Philipp Janser, Daniela Angst, Frédéric Berst, Peter Buehlmayer, Markus Streiff, Christian Beerli, Volker Brinkmann, Danilo Guerini, Paul A Smith, Timothy J Seabrook, Martin Traebert, Klaus Seuwen, René Hersperger, Christian Bruns, Frédéric Bassilana, Marc Bigaud
ABSTRACT
Lymphocyte trafficking is critically regulated by the Sphingosine 1-phosphate receptor-1 (S1P(1)), a G protein-coupled receptor that has been highlighted as a promising therapeutic target in autoimmunity. Fingolimod (FTY720, Gilenya) is a S1P(1) receptor agonist that has recently been approved for the treatment of multiple sclerosis (MS). Here, we report the discovery of NIBR-0213, a potent and selective S1P(1) antagonist that induces long-lasting reduction of peripheral blood lymphocyte counts after oral dosing. NIBR-0213 showed comparable therapeutic efficacy to fingolimod in experimental autoimmune encephalomyelitis (EAE), a model of human MS. These data provide convincing evidence that S1P(1) antagonists are effective in EAE. In addition, the profile of NIBR-0213 makes it an attractive candidate to further study the consequences of S1P(1) receptor antagonism and to differentiate the effects from those of S1P(1) agonists.
Sigma-Aldrich
NIBR-0213, ≥98% (HPLC)
Pricing and availability is not currently available.