MilliporeSigma
  • Realizing Over 13% Efficiency in Green-Solvent-Processed Nonfullerene Organic Solar Cells Enabled by 1,3,4-Thiadiazole-Based Wide-Bandgap Copolymers.
Realizing Over 13% Efficiency in Green-Solvent-Processed Nonfullerene Organic Solar Cells Enabled by 1,3,4-Thiadiazole-Based Wide-Bandgap Copolymers.
Advanced materials (Deerfield Beach, Fla.) (2017-12-07)
Xiaopeng Xu, Ting Yu, Zhaozhao Bi, Wei Ma, Ying Li, Qiang Peng
ABSTRACT
Two novel wide-bandgap copolymers, PBDT-TDZ and PBDTS-TDZ, are developed based on 1,3,4-thiadiazole (TDZ) and benzo[1,2-b:4,5-b']dithiophene (BDT) building blocks. These copolymers exhibit wide bandgaps over 2.07 eV and low-lying highest occupied molecular orbital (HOMO) levels below -5.35 eV, which match well with the typical low-bandgap acceptor of ITIC, resulting in a good complementary absorption from 300 to 900 nm and a low HOMO level offset (≤0.13 eV). Compared to PBDT-TDZ, PBDTS-TDZ with alkylthio side chains exhibits the stronger optical absorption, lower-lying HOMO level, and higher crystallinity. By using a single green solvent of o-xylene, PBDTS-TDZ:ITIC devices exhibit a large open-circuit voltage (V
Sigma-Aldrich
PBDTS-TDZ