MilliporeSigma
  • On the limited role of relA in kanamycin and amino acid starvation induced stringency and subsequent antibiotic cross-protection in Escherichia coli
On the limited role of relA in kanamycin and amino acid starvation induced stringency and subsequent antibiotic cross-protection in Escherichia coli
Journal of Experimental Microbiology and Immunology (2011)
Chiang