MilliporeSigma
  • Recapitulation of metabolic defects in a model of propionic acidemia using patient-derived primary hepatocytes.
Recapitulation of metabolic defects in a model of propionic acidemia using patient-derived primary hepatocytes.
Molecular Genetics and Metabolism (2016)
Chapman KA