MilliporeSigma
  • Association between B7-H1 and cervical cancer: B7-H1 impairs the immune response in human cervical cancer cells.
Association between B7-H1 and cervical cancer: B7-H1 impairs the immune response in human cervical cancer cells.
Experimental and Therapeutic Medicine (2017)
Tao J