MilliporeSigma
  • A validated HPTLC method for determination of ondansetron in combination with omeprazole or rabeprazole in solid dosage form
A validated HPTLC method for determination of ondansetron in combination with omeprazole or rabeprazole in solid dosage form
Indian Journal of Pharmaceutical Sciences (2008)
Raval.B.P, et al.