MilliporeSigma
  • Quantitative amino acid and proteomic analysis: Very low excretion of polypeptides >750 Da in normal urine
Quantitative amino acid and proteomic analysis: Very low excretion of polypeptides >750 Da in normal urine
Kidney International (2004)
Anthony G.W.Norden