MilliporeSigma
  • Study of Physiochemical Parameters for Optimizing Tyrosinase Activity in Rhodococcus ruber C4 and Pseudomonas geniculata C7/C10
Study of Physiochemical Parameters for Optimizing Tyrosinase Activity in Rhodococcus ruber C4 and Pseudomonas geniculata C7/C10
Journal of Pharmacy Research (2015)
Sapna S. Ingle