MilliporeSigma
  • Rho-associated protein kinase 1 (ROCK1) is increased in Alzheimer's disease and ROCK1 depletion reduces amyloid-? levels in brain
Rho-associated protein kinase 1 (ROCK1) is increased in Alzheimer's disease and ROCK1 depletion reduces amyloid-? levels in brain
Journal of Neurochemistry (2016)
Benjamin W