MilliporeSigma
  • Preparation and Evaluation of PEGylated Poly-L-ornithine Complex as a Novel Absorption Enhancer
Preparation and Evaluation of PEGylated Poly-L-ornithine Complex as a Novel Absorption Enhancer
Biologicals : Journal of the International Association of Biological Standardization (2017)
Yusuke Kamiya